MES-Na ≥97.0% (by total nitrogen basis)
Toimittaja: TCI EUROPE
Synonyms:
MES-Na
TCIAM0737-25GEA
85.8
EUR
TCIAM0737-25G
MES-Na ≥97.0% (by total nitrogen basis)
MES sodium salt
Kaava:
C₆H₁₂NNaO₄S MW: 217,22 g/mol |
MDL numero:
MFCD00065473 CAS-numero: 71119-23-8 EINECS: 275-203-2 |
![]() |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...