Isopentyl hexanoate ≥97.0% (by GC) (contains 2-Methylbutyl Hexanoate)
Toimittaja: TCI EUROPE
TCIAH0109-25MLEA
19.8
EUR
TCIAH0109-25ML
Isopentyl hexanoate ≥97.0% (by GC) (contains 2-Methylbutyl Hexanoate)
Isopentyl hexanoate
Kaava:
C₁₁H₂₂O₂ MW: 186,29 g/mol Kiehumispiste: 98°C Leimahduspiste: 88°C |
MDL numero:
MFCD00027280 CAS-numero: 2198-61-0 EINECS: 218-600-8 |
![]() |
Testitulosten erittely
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...