3,4-Dietoksibentsoehappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAD4080-25GEA 42.9 EUR
TCIAD4080-25G
3,4-Dietoksibentsoehappo ≥98.0% (by GC, titration analysis)
3,4-Dietoksibentsoehappo
Kaava: C₁₁H₁₄O₄
MW: 210,23 g/mol
Sulamispiste: 167…168 °C
MDL numero: MFCD00002504
CAS-numero: 5409-31-4
EINECS: 226-478-2
https://fi.vwr.com/stibo/web/std.lang.all/60/27/28986027.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR