HPTLC plates, ProteoChrom®
Toimittaja: Merck
ProteoChrom®
1.05651.0001EA 422 EUR
1.05651.0001 1.05650.0001
HPTLC plates, ProteoChrom®
TLC-levyt
For ProteoChrom® HPTLC silica gel 60 F254s plates, an extra thin layer of high performance silica gel is used, providing highly efficient separation characteristics for 1-D analysis of peptides and protein digests. Up to 20 peptides can be resolved, and as little as 1 to 2 ng per band can be seen.
  • Highly reproducible: Optimised separation and staining procedures
  • Convenient, easy to follow, detailed protocols included
  • Highly stable in water, ideal for use with aqueous solvent systems
For ProteoChrom® HPTLC cellulose sheets, an extra thin layer of optimised microcrystalline cellulose is used. Specially developed protocols for development and staining enable a straightforward 2-D analysis in just four hours.