DL-Arabitol ≥97.0% (by GC)
Toimittaja: TCI EUROPE
TCIAA0517-100MGEA
51.7
EUR
TCIAA0517-100MG
DL-Arabitol ≥97.0% (by GC)
DL-Arabitol
Kaava:
C₅H₁₂O₅ |
MDL numero:
MFCD00070503 CAS-numero: 2152-56-9 EINECS: 218-440-9 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Solubility in Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...