Buffer, HEPPSO, 0,2 M solution, pH 8,0

Toimittaja: Thermo Fisher Scientific
J62529.AKEA 150 EUR
J62529.AK
Buffer, HEPPSO, 0,2 M solution, pH 8,0
Biokemialliset puskuriliuokset Biological Buffers
CAS No.: 68399-78-0
Physical form: liquid
MDL No.: MFCD00038351
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR