(S)-(-)-3-Acetamidopyrrolidine ≥98.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAA1170-5GEA
157
EUR
TCIAA1170-5G
(S)-(-)-3-Acetamidopyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-3-Acetamidopyrrolidine
Kaava:
C₆H₁₂N₂O MW: 128,17 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD00082748 CAS-numero: 114636-31-6 |
Testitulosten erittely
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -46.0 to -51.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...