D-(+)-Allose

Toimittaja: Molekula

MOLE50241498-250MGEA 143 EUR
MOLE50241498-250MG
D-(+)-Allose
D-(+)-Allose
Kaava: C₆H₁₂O₆
MW: 180,1559 g/mol
MDL numero: MFCD00135833
CAS-numero: 2595-97-3
EINECS: 219-994-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR