L(-)-Talose ≥98.0% (by HPLC)

Toimittaja: TCI EUROPE

TCIAT1767-100MGEA 401 EUR
TCIAT1767-100MG
L(-)-Talose ≥98.0% (by HPLC)
L(-)-Talose
Kaava: C₆H₁₂O₆
MDL numero: MFCD00135850
CAS-numero: 23567-25-1
EINECS: 245-744-9
https://fi.vwr.com/stibo/web/std.lang.all/57/43/28995743.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -17.0 to -23.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR