Sodium-2-aminonaphthalene-1-sulphonate ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI EUROPE
Varoitus

TCIAA0352-5GEA 23.1 EUR
TCIAA0352-5G
Sodium-2-aminonaphthalene-1-sulphonate ≥98.0% (by HPLC, total nitrogen)
Sodium-2-aminonaphthalene-1-sulphonate
Kaava: C₁₀H₈NNaO₃S
MDL numero: MFCD00070484
CAS-numero: 25293-52-1
EINECS: 246-793-9

Order Now

Testitulosten erittely

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR