Lämpömittari, radiosignaalitoiminen langaton lämpömittari, Traceable®
Toimittaja: VWR Collection
620-1982EA 212 EUR
VWRI620-1982
Lämpömittari, radiosignaalitoiminen langaton lämpömittari, Traceable®
This radio signal thermometer can read refrigerator temperatures from up to 100 feet away. They can provide readings from up to three remote refrigerator sensors, to a main unit (frequency of 433 MHz).
  • Distinctive alarm sounds when temperature rises above/falls below high and low set points, set in 1 °C increments
  • Sealed GRAS (Generally Recognized as Safe by FDA) bottle insulated from rapid temperature changes
  • Easy-to-read two lines display: Data update every 30 seconds, minimum/maximum memories
Sertifikaatit: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Toimitustiedot: Supplied with a main unit, a remote unit, batteries, flip-open stand, wall mount, VELCRO® and Traceable® calibration certificate.