4-Phenyl-1-buten-4-ol ≥97.0% (by GC)
Toimittaja: TCI EUROPE
TCIAP1329-5GEA
175
EUR
TCIAP1329-5G
4-Phenyl-1-buten-4-ol ≥97.0% (by GC)
4-Phenyl-1-buten-4-ol
Kaava:
C₁₀H₁₂O MW: 148,20 g/mol Leimahduspiste: 132°C |
MDL numero:
MFCD00039617 CAS-numero: 936-58-3 |
Testitulosten erittely
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...