Lead standard solution (0.1% Pb) Reag. Ph. Eur. 5001700
Toimittaja: VWR Chemicals
88087.180EA 91.9 EUR
88087.180
Lead standard solution (0.1% Pb) Reag. Ph. Eur. 5001700
Lead standard solution (0.1% Pb)
Tilaustiedot: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Order Now
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022