VWR® ARISTAR®, Multi-Element Quality Control Standard 1000 mg/L, 21 Elements in Nitric Acid 4% for ICP
Toimittaja: Avantor
VWR® ARISTAR®
87629.180EA 592 EUR
87629.180
VWR® ARISTAR®, Multi-Element Quality Control Standard 1000 mg/L, 21 Elements in Nitric Acid 4% for ICP
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards
21 elements 1000 mg/L: Ag, As, Al, B, Ba, Bi, Cd, Co, Cr, Cu, Fe, Ga, In, Li, Mn, Ni, Pb, Si, Sr, Tl, Zn.
Sertifikaatit: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Toimitustiedot: Supplied with certificate of analysis. The Si standard is supplied in a separate 100 ml bottle.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022