Flame-retardant bomber jacket, high visibility, Delano 9485
Toimittaja: Sioen
Sio-Start® FR
SIOE9485/278/MEA 253 EUR
SIOE9485/278/M SIOE9485/278/3XL SIOE9485/278/L SIOE9485/278/S SIOE9485/278/1XL SIOE9485/278/2XL
Flame-retardant bomber jacket, high visibility, Delano 9485
Työvaatteet
Anti-static jacket made of Sio-Start FR AST fabric: 100% polyester fabric with 100% FR PU coating + AST. With fixed flame-retardant cotton lining and detachable flame-retardant fleece lining in body (without sleeves, can not be worn separately).
  • Highly breathable, water- and windproof, high tear resistance
  • Straight collar with foldaway hood
  • Zip closure under flap with touch & close fastening
  • Two patched pockets, Napoleon pocket, to inside pockets in linings
  • Round inset sleeves, sleeve narrowing by press studs
  • Elastic drawstring in hem
The fabric is water repellent on the outside and features a moisture attracting coating on the inside.
Back length: 75 cm
Weight: 250 g/m²
Sertifikaatit: EN 13034 (Type PB/6), EN 531 (A/B2/C1), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 343 (3-3), EN 471 (3-2)
Detachable lining only: EN 14058 (1XXXX), EN ISO 14116 (Index 1)