Industrial jackets, CI, design C, 490L PR54
Toimittaja: Fristads Kansas Group
KWIN100537940XSEA 0 EUR
KWIN100537940XS KWIN100537940S KWIN100537940L KWIN100537940XL KWIN100537940M KWIN1005379402XL
Industrial jackets, CI, design C, 490L PR54
Työvaatteet
Functional jacket made of 65% polyester, 35% cotton, with soft nap on reverse side.
  • Zip closure
  • Two chest pockets, one double with flap and velcro closure
  • Two front pockets, glove pocket, sleeve pocket, concealed ID-card pocket, concealed pen pocket
  • Mobile phone pocket with loop for earpiece, two inside pockets
  • Adjustable waist, rounded elbows, adjustable cuffs
  • OEKO-TEX® certified
Weight: 260 g/m²
Please note: These jackets can be customised. You can chose between four base colours and 12 accent and piping colours to suit your company's profile. Please contact your local Avantor customer service team for further information.