Model of brain stem in 8 parts
Toimittaja: Somso
SOMMBS25/2EA 0 EUR
SOMMBS25/2
Model of brain stem in 8 parts
Mallit Ihmisbiologia
After Prof. Dr. Dr. J. W. Rohen, Department of Anatomy of the University Erlangen. The model separates into 8 parts as follows: brain stem (2), corpus striatum, insula (2), nucleus lentiformis (left), internat capsule (right) and ventricles of the brain.
  • Natural size
  • Made in SOMSO-Plast®
  • On a stand with green base
H×W×D: 16×12×12 cm
Weight: 0,38 kg