3-(Trimethylsilylethynyl)pyridine ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAT1963-1GEA
161
EUR
TCIAT1963-1G
3-(Trimethylsilylethynyl)pyridine ≥98.0% (by GC)
3-(Trimethylsilylethynyl)pyridine
Kaava:
C₁₀H₁₃NSi MW: 175,31 g/mol Leimahduspiste: 94°C Storage Temperature: Pakastin |
MDL numero:
MFCD01863744 CAS-numero: 80673-00-3 |
Testitulosten erittely
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...