5-Benzyl-1H-tetrazole

Toimittaja: Molekula

MOLEM55454207EA 0 EUR
MOLEM55454207
5-Benzyl-1H-tetrazole
5-Benzyl-1H-tetrazole
Kaava: C₈H₈N₄
MW: 160,18 g/mol
MDL numero: MFCD00068730
CAS-numero: 18489-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR