2-(Methylthio)benzimidazole ≥98%

A10686.09EA 86.4 EUR
A10686.09
2-(Methylthio)benzimidazole ≥98%
2-(Methylthio)benzimidazole
Kaava: C₈H₈N₂S
Sulamispiste: 203…205 °C
MDL numero: MFCD00005594
CAS-numero: 7152-24-1
EINECS: 230-494-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR