Pyridazine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP1907-1GEA
139
EUR
TCIAP1907-1G
Pyridazine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Pyridazine-4-carboxylic acid
Kaava:
C₅H₄N₂O₂ MW: 124,10 g/mol Sulamispiste: 238°C |
MDL numero:
MFCD00011576 CAS-numero: 50681-25-9 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...