3-(Hydroxymethyl)-5-methylisoxazole

Toimittaja: Thermo Scientific

MAYBCC10409DAEA 0 EUR
MAYBCC10409DA
3-(Hydroxymethyl)-5-methylisoxazole
3-(Hydroxymethyl)-5-methylisoxazole
Kaava: C₅H₇NO₂
MW: 113,11 g/mol
MDL numero: MFCD00085129
CAS-numero: 35166-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR