4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester 95%

H52270.03EA 157 EUR
H52270.03
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester 95%
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester
Kaava: C₁₇H₂₇BN₂O₃
MW: 318,22 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05864307
CAS-numero: 832114-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR