2-Cyanobenzamide ≥98%

Varoitus

B21596.06EA 624 EUR
B21596.06
2-Cyanobenzamide ≥98%
2-Cyanobenzamide
Kaava: C₈H₆N₂O
MW: 146,15 g/mol
Sulamispiste: 172…173 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014797
CAS-numero: 17174-98-0
EINECS: 241-224-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR