4-(Dimethylamino)thiophenol ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAD4047-1GEA 88 EUR
TCIAD4047-1G
4-(Dimethylamino)thiophenol ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)thiophenol
Kaava: C₈H₁₁NS
Kiehumispiste: 260°C
MDL numero: MFCD01658123
CAS-numero: 4946-22-9
https://fi.vwr.com/stibo/web/std.lang.all/59/94/28985994.jpg

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR