Diphenylzinc ≥98%

Varoitus

87908.06EA 1570 EUR
87908.06
Diphenylzinc ≥98%
Diphenylzinc
Kaava: C₁₂H₁₀Zn
MW: 219,6 g/mol
Kiehumispiste: 280…285 °C
Sulamispiste: 107 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014071
CAS-numero: 1078-58-6
EINECS: 214-082-2
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR