2-Methylisonicotinic acid ≥96%

Varoitus

H57969.03EA 225 EUR
H57969.03
2-Methylisonicotinic acid ≥96%
2-Methylisonicotinic acid
Kaava: C₇H₇NO₂
MW: 137,14 g/mol
Sulamispiste: 295…300 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00513435
CAS-numero: 4021-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR