8-(Bromomethyl)quinoline

Toimittaja: Molekula

MOLE12992569-1GEA 0 EUR
MOLE12992569-1G
8-(Bromomethyl)quinoline
8-(Bromomethyl)quinoline
Kaava: C₁₀H₈BrN
MW: 222,0800 g/mol
MDL numero: MFCD00047621
CAS-numero: 7496-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR