4-Undecanol ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAU0038-5MLEA 141 EUR
TCIAU0038-5ML
4-Undecanol ≥98.0% (by GC)
4-Undecanol
Kaava: C₁₁H₂₄O
Kiehumispiste: 80°C
Leimahduspiste: 66°C
MDL numero: MFCD00046724
CAS-numero: 4272-06-4
EINECS: 224-264-3
https://fi.vwr.com/stibo/web/std.lang.all/71/99/28997199.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR