1-Bromo-3,5-difluoro-4-iodobenzene ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB3058-5GEA 101 EUR
TCIAB3058-5G
1-Bromo-3,5-difluoro-4-iodobenzene ≥97.0% (by GC)
1-Bromo-3,5-difluoro-4-iodobenzene
Kaava: C₆H₂BrF₂I
MW: 318,8854 g/mol
MDL numero: MFCD01861126
CAS-numero: 160976-02-3
https://fi.vwr.com/stibo/web/std.lang.all/45/01/28934501.jpg

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR