Flame-retardant jackets, Nomex® Comfort, 4821 NX
Toimittaja: Fristads Kansas Group
KWIN1009296762XLEA 0 EUR
KWIN1009296762XL KWIN1009296763XL KWIN100929676L KWIN100929676S KWIN100929676M KWIN1009296764XL
Flame-retardant jackets, Nomex® Comfort, 4821 NX
Työvaatteet
Protective jacket made of inherent Nomex® Comfort, dope dyed. The fabric consists of 93% Meta-Aramid, 5% Para-Aramid, 2% anti-static fibre. Provides protection against open flames and chemicals. When exposed to intense heat, the fabric carbonises and thickens, forming a virtually impenetrable barrier.
  • Front placket opening with zip to top of collar and concealed snap fastening
  • Two chest pockets with flaps and concealed snap fastening
  • Sleeve pocket with flap and concealed snap fastening
  • Inside pocket and mobile phone pocket with velcro® fastening
  • Two front pockets with self-closing flap
  • Adjustable waist and cuffs
Weight: 260 g/m²
Sertifikaatit: EN 531 (class A, B1 and C1), EN 1149-5, EN 13034 (type BP[6])