Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III) ≥97.0% (by titrimetric analysis) NMR shift reagent
Toimittaja: TCI EUROPE
TCIAT1511-1GEA
94.6
EUR
TCIAT1511-1G
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III) ≥97.0% (by titrimetric analysis) NMR shift reagent
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III)
Kaava:
C₃₀H₃₀F₂₁O₆Pr MW: 1026,43 g/mol |
MDL numero:
MFCD00064656 CAS-numero: 17978-77-7 EINECS: 241-902-6 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...