4-Cyano-4'-methylbiphenyl ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAC1741-25GEA
179
EUR
TCIAC1741-25G
4-Cyano-4'-methylbiphenyl ≥98.0% (by GC)
4-Cyano-4'-methylbiphenyl
Kaava:
C₁₄H₁₁N MW: 193,2438 g/mol |
MDL numero:
MFCD03703444 CAS-numero: 50670-50-3 EINECS: 256-701-9 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...