4-(2,2,2-Trifluoroethoxy)phenylboronic acid ≥98%

Varoitus

H52889.MDEA 356 EUR
H52889.MD H52889.03
4-(2,2,2-Trifluoroethoxy)phenylboronic acid ≥98%
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
Kaava: C₈H₈BF₃O₃
MW: 219,95 g/mol
Sulamispiste: 191…194 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09027239
CAS-numero: 886536-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR