2'-(N,N-Dimethylamino)-5'-nitroacetanilide ≥98.0% (by titrimetric analysis)
Toimittaja: TCI EUROPE
TCIAD2020-1GEA
137
EUR
TCIAD2020-1G
2'-(N,N-Dimethylamino)-5'-nitroacetanilide ≥98.0% (by titrimetric analysis)
2'-(N,N-Dimethylamino)-5'-nitroacetanilide
Kaava:
C₁₀H₁₃N₃O₃ |
MDL numero:
MFCD00130122 CAS-numero: 5367-36-2 |
Testitulosten erittely
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 164.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...