Azocyclotin

Toimittaja: EHRENSTORFER GMBH

EHERC10400000EA 131 EUR
EHERC10400000
Azocyclotin
Azocyclotin
Kaava: C₂₀H₃₅N₃Sn
MDL numero: MFCD00055290
CAS-numero: 41083-11-8
EINECS: 255-209-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR