Bathocuproin sulphonate disodium salt

Toimittaja: Molekula

MOLEM15611838EA 137 EUR
MOLEM15611838
Bathocuproin sulphonate disodium salt
Bathocuproin sulphonate disodium salt
Kaava: C₂₆H₁₈N₂Na₂O₆S₂
MDL numero: MFCD00011679
CAS-numero: 52698-84-7
EINECS: 258-111-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR