N(α)-Boc-L-tryptophan

Toimittaja: Molekula

Synonyms: Boc-Trp-OH

MOLEM25104502EA 94.3 EUR
MOLEM25104502
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan
Kaava: C₁₆H₂₀N₂O₄
MW: 304,3 g/mol
MDL numero: MFCD00065595
CAS-numero: 13139-14-5
EINECS: 236-072-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR