1-Phenyl-1-(trimethylsilyloxy)ethylene ≥95.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP1280-10MLEA
138
EUR
TCIAP1280-10ML
1-Phenyl-1-(trimethylsilyloxy)ethylene ≥95.0% (by GC)
1-Phenyl-1-(trimethylsilyloxy)ethylene
Kaava:
C₁₁H₁₆OSi MW: 192,34 g/mol Kiehumispiste: 94°C Leimahduspiste: 78°C |
MDL numero:
MFCD00008582 CAS-numero: 13735-81-4 EINECS: 237-308-1 |
Testitulosten erittely
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...