4-Methoxyazobenzene ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAP0142-25GEA 211 EUR
TCIAP0142-25G
4-Methoxyazobenzene ≥98.0% (by GC)
4-Methoxyazobenzene
Kaava: C₁₃H₁₂N₂O
Kiehumispiste: 340°C
MDL numero: MFCD00039804
CAS-numero: 2396-60-3
EINECS: 219-250-9
https://fi.vwr.com/stibo/web/std.lang.all/08/56/28990856.jpg

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR