Nordihydroguaiaretic acid, white powder
Toimittaja: MP Biomedicals
|
Varoitus
|
Synonyms:
4,4'-(2,3-Dimethylbutane-1,4-diyl)bis(benzene-1,2-diol)
ICNA0215594490EA
48.9
EUR
ICNA0215594490
Nordihydroguaiaretic acid, white powder
Nordihydroguaiaretic acid
Inhibitor of lipoxygenases
Soluble in ethanol (5% w/v), methanol, ether, glycol, PEG; insoluble in water
Varoitus: Research Use Only (RUO).
Kaava:
C₁₈H₂₂O₄ MW: 302,37 g/mol Storage Temperature: Pakastin |
MDL numero:
MFCD00002206 CAS-numero: 500-38-9 EINECS: 207-903-0 |
Testitulosten erittely
Identity Test | Passes |
Melting Point | 182 - 192 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...