Chromium(III) picolinate

Toimittaja: U.S. Pharmacopeia

USPH1133638EA 302 EUR
USPH1133638
Chromium(III) picolinate
Chromium(III) picolinate
Kaava: C₁₈H₁₂CrN₃O₆
MDL numero: MFCD00068715
CAS-numero: 14639-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR