2-Cyano-4'-methylbiphenyl ≥99.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAC1446-10GEA
41.8
EUR
TCIAC1446-10G
2-Cyano-4'-methylbiphenyl ≥99.0% (by GC)
2-Cyano-4'-methylbiphenyl
Kaava:
C₁₄H₁₁N MW: 193,26 g/mol |
MDL numero:
MFCD00151805 CAS-numero: 114772-53-1 EINECS: 422-310-9 |
Testitulosten erittely
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 50.0 to 54.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...