α-Phenylcinnamic acid ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAP1300-25GEA 292 EUR
TCIAP1300-25G
α-Phenylcinnamic acid ≥98.0% (by titrimetric analysis)
α-Phenylcinnamic acid
Kaava: C₁₅H₁₂O₂
MDL numero: MFCD00004252
CAS-numero: 91-48-5
EINECS: 202-069-4
https://fi.vwr.com/stibo/web/std.lang.all/19/46/28991946.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR