4-Chlorodiphenyl sulphone ≥96.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAC1451-25GEA
136
EUR
TCIAC1451-25G
4-Chlorodiphenyl sulphone ≥96.0% (by GC)
4-Chlorodiphenyl sulphone
Kaava:
C₁₂H₉ClO₂S Kiehumispiste: 389°C |
MDL numero:
MFCD00007549 CAS-numero: 80-00-2 EINECS: 201-242-1 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 91.0 to 94.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...