4-Phenyl-1-butyne ≥97.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP0358-5MLEA
92.4
EUR
TCIAP0358-5ML
4-Phenyl-1-butyne ≥97.0% (by GC)
4-Phenyl-1-butyne
Kaava:
C₁₀H₁₀ MW: 130,19 g/mol Kiehumispiste: 190°C Leimahduspiste: 66°C |
MDL numero:
MFCD00041667 CAS-numero: 16520-62-0 |
Testitulosten erittely
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...