Diethoxymethylsilane ≥95.0% (by GC) hydrosilylating reagent
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAD2403-25MLEA
82.5
EUR
TCIAD2403-25ML
Diethoxymethylsilane ≥95.0% (by GC) hydrosilylating reagent
Diethoxymethylsilane
Kaava:
C₅H₁₄O₂Si MW: 134,25 g/mol Kiehumispiste: 97°C Leimahduspiste: 10°C |
MDL numero:
MFCD00026749 CAS-numero: 2031-62-1 EINECS: 217-982-3 UN: 1993 ADR: 3,II |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...