Diethoxymethylvinylsilane ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAD2318-25MLEA 27.5 EUR
TCIAD2318-25ML
Diethoxymethylvinylsilane ≥97.0% (by GC)
Diethoxymethylvinylsilane
Kaava: C₇H₁₆O₂Si
MW: 160,29
Kiehumispiste: 134°C
Leimahduspiste: 17°C
MDL numero: MFCD00009067
CAS-numero: 5507-44-8
EINECS: 226-850-4
UN: 1993
ADR: 3,II
https://fi.vwr.com/stibo/web/std.lang.all/92/51/28979251.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR