2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole ≥99%

L07874.03EA 165 EUR
L07874.03
2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole ≥99%
2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole
Kaava: C₂₆H₁₈N₂O
MW: 374,45 g/mol
Sulamispiste: 235…238 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00042666
CAS-numero: 2043-06-3
EINECS: 218-046-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR