These disposable wipes are made of 100% Vileda Professional polyester fibre. They are ideal for standard cleanroom cleaning applications.
- Optimum stitch density for specific applications
- Cleanroom laundered and packed
- Sealed edges via laser cutting for low particle release
PolyControl 400: for ISO class 4 - 9 environments
- Rough surface wipe
- Ultra low level of particles and extractables
- Resists abrasion under rigorous use
PolyControl 600: for ISO class 5 - 9 environments
- Delicate surface wipes
- Low level of particles and extractables
- Fine polyester fibres with high absorbency
PolyControl 900: for ISO class 4 - 9 environments
- High absorbency and spill pick-up wipes
- Ultra low level of particles and extractables
- Unique two-ply construction for excellent absorbency
Colour: White